missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ High Visibility Barrier Backed Safety Mat Roll

Combine safety signage with absorbency. The Brady™ High Visibility Barrier Backed Series Roll is the perfect option for high traffic walkways and foot paths. The bright color and printed message bring awareness while the absorbency prevents slips, trips and falls.

$168.40 - $168.40

Specifications

Absorbancy 11.7 gl/gs
Color Black on yellow
Length (English) 100 ft.
For Use With (Application) High traffic areas where the risk for slips, trips and falls is greatest
Material Polypropylene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W) Width (English) Capacity (English) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Width (English) Capacity (English) Price Quantity  
19-103-105 Brady™
CHBB15
100 ft. x 15 in. 15 in. 13.4 gal. Each for $168.40
Specifications

Description

  • Great addition to your absorbent program
  • Combines absorbency, durability and safety signage into one versatile product
  • Designed as an all-purpose absorbent for industrial applications, this mat is the perfect option for high-traffic walkways and foot paths
  • Barrer backing resists tearing and helps keep the mat in place
  • Bright color and printed message bring attention to areas where there is a high risk of slips, trips and falls
Description & Specifications

Specifications

11.7 gl/gs
100 ft.
Polypropylene
Black on yellow
High traffic areas where the risk for slips, trips and falls is greatest
1