Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles
Designed to fit most Soxhlet extraction units. Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles are made of 100% borosilicate glass microfiber. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
$238.63 - $458.91
Specifications
Material | Borosilicate glass fibers |
---|---|
For Use With (Application) | Smoke stack gas monitoring. Soxhlet extraction. Analyzing pesticide residues. Determining oil/fat content of foods. Analysis of oil and grease in solid wastes. |
Max. Temperature (Metric) | 500°C |
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) Inside | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) Inside | Price | Quantity | ||||
09-657-1A
|
Cytiva (Formerly GE Healthcare Life Sciences)
2814199 |
90 mm | 19 mm |
Pack of 25 Each for $238.63
|
|
09-657-1C
|
Cytiva (Formerly GE Healthcare Life Sciences)
2814300 |
100 mm | 30 mm |
Pack of 25 Each for $394.74
|
|
09-657-1B
|
Cytiva (Formerly GE Healthcare Life Sciences)
2814432 |
123 mm | 43 mm |
Pack of 25 Each for $458.91
|
|
Description
Designed to fit most Soxhlet extraction units. Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles are made of 100% borosilicate glass microfiber. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
- Resistant to temperatures up to 500°C
- Binder-free, no chemical additives
- Designed to fit most commercially available Soxhlet extractors.
- Effective when used with solvents incompatible with cellulose materials
- Applications: analysis of hot and acidic gases during emission control and smoke stack gas monitoring, analysis of pesticide residues using Soxhlet extraction, determination of oil/fat content of foods, analysis of oil and grease in solid wastes>
Specifications
Borosilicate glass fibers | |
Smoke stack gas monitoring. Soxhlet extraction. Analyzing pesticide residues. Determining oil/fat content of foods. Analysis of oil and grease in solid wastes. | |
500°C |