Please login to your online account to display your discounted pricing
High-Frequency Tesla Coil
Safe source of high-frequency, high-voltage electricity.
Manufacturer: fisher scientific  10205000
 View more versions of this product
Catalog No. S43505
  • / Each
Description & Specifications
Specifications
Includes Vibrator, two capacitors, cord and plug, primary coil, secondary coil with 400 turns of wire, Plastic housing, lamp socket, two radiating antenna plates Neon bulb, discharge electrode, Plastic and Copper wires, instructions with 19 experiments
Product Type Tesla coil
Voltage 110V
For Use With (Application) Transmit electricity without wires; Demonstrates fundamental principles of high frequency electrical phenomena
Accessories
Accessories for Catalog Number: S43505
Item Mfr. No. Item Description Price Per Unit Quantity  
S68853
High-Frequency Tesla Coil
fisher scientific
10-212
Safe source of high-frequency, high-voltage electricity. Each for $18.35
S94213
Neon Wand
fisher science education
10-213
Demonstrate the effects of ionization and excitation of different gases. Each for $9.15