Please login to your online account to display your discounted pricing

High Five™ Spunbond Polypropylene Shoe Covers with Tread

Sanitary, with 2.5 in. tacky strip on sole to keep wearer from slipping

$35.54 - $37.62

Specifications

Closure Type Elastic
Color Blue
Material Polypropylene
Resistance Abrasion
For Use With (Application) Ideal to provide protection against general splash and spills
View More Specs
Products
Catalog Number Mfr. No. Includes Size Price Quantity    

19-167-939

 
high five™
AS613
2.5 in. blue and white tread w/ tacky strip Universal Case of 300 Each for $37.62

19-167-942

 
high five™
AS624
2.5 in. blue and white tread w/ tacky strip X-Large Case of 300 Each for $35.54
Description & Specifications

Specifications

Closure Type Elastic
Color Blue
Material Polypropylene
Resistance Abrasion
For Use With (Application) Ideal to provide protection against general splash and spills

  • Strong, abrasion-resistant spunbond polypropylene
  • Full elastic top stretches to allow for easy on and off
  • Blue with white tread

Bulk pack: 300/case; dispenser pack: three dispensers of 100.