High Five™ Spunbond Polypropylene Lab Coats
Economical protection from dust, dirt and grime
$41.76 - $42.21
Specifications
Closure Type Snap
Construction Breathable Construction Keeps Workers Cooler
Material Spunbond Polypropylene
For Use With (Application) Protection from dust, dirt and grime
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Size Price Quantity  
Catalog Number Mfr. No. Color Size Price Quantity  
19-167-930 High Five™
AL602
White Medium Case of 30 Each for $42.21
19-167-931 High Five™
AL603
White Large Case of 30 Each for $42.21
19-167-932 High Five™
AL604
White X-Large Case of 30 Each for $42.21
19-167-933 High Five™
AL605
White 2X-Large Case of 30 Each for $41.76
Specifications
Description
  • Breathable construction keeps workers cooler
  • No pockets to catch contaminants
  • Elastic wrists
  • Four-snap closure
  • White
Description & Specifications
Specifications
Snap
Breathable Construction Keeps Workers Cooler
Spunbond Polypropylene
Protection from dust, dirt and grime