Please login to your online account to display your discounted pricing

High Five™ Nitrile Powder-Free Exam Gloves

An economical alternative to latex exam gloves featuring texturing for enhanced gripping power

$8.55 - $85.54

Specifications

Color Blue
Length (English) 9.5 in.
Length (Metric) 24.13cm
Material Nitrile
Sterility Non-sterile
View More Specs
Products
Catalog Number Mfr. No. Certifications Size Price Quantity    

19-167-08

 
microflex corporation
N291
AQL 1.5, ISO9001 QMS Small Case of 1000 Each for $85.54

19-167-09

 
microflex corporation
N292
AQL 1.5, ISO9001 QMS Medium Pack of 100 Each for $8.55
Case of 10 Pack for $81.80

19-167-10

 
microflex corporation
N293
AQL 1.5, ISO9001 QMS Large Case of 1000 Each for $85.54

19-167-11

 
microflex corporation
N294
AQL 1.5, ISO9001 QMS X-Large Case of 1000 Each for $85.54
Description & Specifications

Specifications

Color Blue
Length (English) 9.5 in.
Length (Metric) 24.13cm
Material Nitrile
Sterility Non-sterile
Disposable Yes
Glove Cuff Style Beaded/Rolled
Grade Medical, exam
Hand Ambidextrous
Latex Free Yes
Tensile Strength 20.5
Thickness (English) 3 mil

  • Soft, stretchy formulation for durability and comfort
  • Length: 9.5 in.
  • Palm: 3mil for enhanced tactile sensitivity
  • Medical/exam grade
  • Blue

510K approved for medical use