Please login to your online account to display your discounted pricing

High Five™ Microporous Lab Coats

Breathability with durability

$72.67 - $84.79

Specifications

Closure Type Snap
Color White
Material Microporous Film
No. of Pockets 2
For Use With (Application) General purpose protection from dirt, grime, splashes and spills
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-167-922

 
high five™
AL244
X-Large Case of 30 Each for $78.72

19-167-923

 
high five™
AL245
2X-Large Case of 30 Each for $81.76

19-167-924

 
high five™
AL246
3X-Large Case of 30 Each for $84.79

19-167-921

 
high five™
AL243
Large Case of 30 Each for $75.71

19-167-920

 
high five™
AL242
Medium Case of 30 Each for $72.67
Description & Specifications

Specifications

Closure Type Snap
Color White
Material Microporous Film
No. of Pockets 2
For Use With (Application) General purpose protection from dirt, grime, splashes and spills

  • Cloth-like, microporous material
  • General purpose protection from dirt, grime, splashes and spills
  • Economical and lightweight
  • Low linting and antistatic
  • Elastic wrists, five-snap closure and two pockets
  • White