Thermo Scientific™ High-Capacity Section Dryer
Work with a large drying area while using minimal bench space with the Thermo Scientific™ High-Capacity Section Dryer.
$8,459.40 - $8,459.40
Specifications
Chamber Material Stainless steel
Temperature Control Range 40 to 90°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Voltage Price Quantity  
Catalog Number Mfr. No. Voltage Price Quantity  
28-618-207 Thermo Scientific™
SHB3120202
120V Each for $8,459.40
Specifications
Description
Work with a large drying area while using minimal bench space with the Thermo Scientific™ High-Capacity Section Dryer. Available in 120V and 230V versions.Thermo Scientific High-Capacity Section Dryer facilitates accurate and constant temperature control from 40 to 90ºC.
  • Digital controller facilitates accurate and constant temperature control from 40° to 90°C.
  • Polished stainless steel drying chamber includes an autoclavable drip tray.
  • Includes air intake with removable filters and easy-to-clean epoxy-coated external finish.
Description & Specifications
Specifications
Stainless steel
40 to 90°C