Please login to your online account to display your discounted pricing

Thermo Scientific™ HIF-1α (Hypoxia-Inducible Factor-1α) Ab-1, Mouse Monoclonal Antibody


Recommended for Immunofluorescence and Gel Supershift, Thermo Scientific™ HIF-1α (Hypoxia-Inducible Factor-1α) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™  MS701P1ABX

 View more versions of this product

Catalog No. MS701P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Applications Immunofluorescence
Applications Western Blotting
Quantity 100µL
Regulatory Status RUO
Clone OZ12
Cross Reactivity Human
Antigen HIF-1alpha (Hypoxia-Inducible Factor-1alpha) Ab-1
Format Concentrated
Immunogen hHIF-1alpha (aa 530-826) protein
Isotype IgG2a kappa
Research Discipline Stress Response
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Classification Monoclonal

Recommended for Immunofluorescence and Gel Supershift, Thermo Scientific™ HIF-1α (Hypoxia-Inducible Factor-1α) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

HIF-1 (hypoxia-inducible factor-1) is a heterodimer consisting of an alpha and a β subunit, both belonging to the basic-helix-loop-helix Per-aryl hydrocarbon receptor nuclear translocator-Sim (PAS) family of transcription factors. HIF-1 is a key component of a widely operative transcriptional response activated by hypoxia, cobaltous ions, and iron chelation. HIF-1 activates transcription of hypoxia-inducible genes, including those encoding: erythropoietin, vascular endothelial growth (VEGF), heme oxygenase-1, inducible nitric oxide synthase, and the glycolytic enzymes aldolase A, enolase 1, lactate dehydrogenase A, phosphofructokinase I, and phosphoglycerate kinase 1. The C-terminal of HIF-1 alpha binds to p300. p300/CBP-HIF complexes participate in the induction of hypoxia-responsive genes, including VEGF.

Host Species: Mouse

Clone: OZ12

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 530-826

Immunogen: hHIF-1α (aa 530-826) protein

Molecular Weight: 120kDa

Positive Control: Mammalian cells cultured under reduced O2 tension

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO