Please login to your online account to display your discounted pricing

HGAL Mouse anti-Human, Clone: 1H1-A7, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $368.30

Specifications

Antigen HGAL
Clone 1H1-A7
Host Species Mouse
Gene Alias Human germinal center-associated lymphoma, GCET2
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122829

 
life technologies
LS14975880
25μg Each for $146.45

501122830

 
life technologies
LS14975882
100μg Each for $368.30
Description & Specifications

Specifications

Antigen HGAL
Clone 1H1-A7
Host Species Mouse
Gene Alias Human germinal center-associated lymphoma, GCET2
Species Reactivity Human, Mouse
Applications Enzyme Linked Immunosorbent Assay
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunoprecipitation
Applications Microscopy
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 1H1-A7 recognizes human germinal center-associated lymphoma (HGAL) protein. HGAL is expressed in germinal centers, spleen, thymus, and germinal center-derived lymphomas. HGAL is localized to the cytoplasm of cells within the dark zone of the germinal center and functions to enhance the activation of RhoA leading to negative regulation of lymphocyte motility. HGAL expression has been used to help elucidate nodal marginal zone lymphoma (NMZL) from cases of diffuse follicle center lymphoma. Additionally, HGAL expression was shown to correlate with survival in patients with diffuse large B-cell lymphoma (DLBCL).

The 1H1-A7 antibody recognizes the protein product of the M17 gene, the mouse homolog of the HGAL protein.