HF Scientific™ ProCal™ Turbidity Standard Reagents Kit for Hach™ 2100Q Portable Turbidimeter
Standards not harmful to the environment or to worker safety
Manufacturer:  HF Scientific™ HF52500
 View more versions of this product
Catalog No. 15-393-636
Description
Description
  • Pre-diluted
  • Do not include formazin
  • Preservative used enhances stability
  • Biodegradable, safe and non-toxic
  • Ready and easy to use
  • Factory certified by HF scientific
  • Custom NTU produced
  • Available in different size bottles
Specifications
Specifications
500mL of each 0.0, 20, 100, 800 NTU value
EPA
16.9 oz.
500mL
Primary Turbidity Standard Kit
For Hach™ 2100Q portable turbidimeter
0, 20, 100, 800 NTU
Documents