HF Scientific™ ProCal™ Primary Turbidity Standards for Hach™ 2100N Laboratory Turbidimeter
Standards not harmful to the environment or to worker safety
Manufacturer:  HF Scientific™ HF52200
 View more versions of this product
Catalog No. 15-393-606
Description
Description
  • Pre-diluted
  • Do not include formazin
  • Preservative used enhances stability
  • Biodegradable, safe and non-toxic
  • Ready and easy to use
  • Factory certified by HF scientific
  • Custom NTU produced
  • Available in different size bottles
Specifications
Specifications
Primary Turbidity Standard
For Hach™ 2100N turbidimeter
200 NTU
EPA
8.45 oz.
250mL
Documents