missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HF Scientific™ Powder Pop™ DPD Reagent for Chlorine Testing

For testing free or total chlorine

Manufacturer:  HF Scientific™ HF10303

 View more versions of this product

Catalog No. 15-393-59



Description

Description

  • Deliver doses of DPD reagent measured for 5, 10 or 25mL (0.2, 0.3 or 0.8 oz.) testing samples
  • Individually packaged

Specifications

Specifications

200/Pk.
For testing total chlorine
5mL
DPD Reagent
0.16 oz.
Documents