Please login to your online account to display your discounted pricing
HF scientific Powder Pop™ Dispensers for Chlorine Testing
For testing free or total chlorine
$56.34 - $66.70
Specifications
Product Type Dispenser
No. of Tests 100.0
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Sample Size Price Quantity    
36-099-1473
hf scientific
HF10500
For testing free chlorine HF scientific and Hach™ photometers 5mL Each for $56.34
15-393-152
hf scientific
HF10504
For testing free chlorine Hach™ and Hanna photometers 10mL Each for $66.70
15-393-153
hf scientific
HF10506
For testing total chlorine Hach™ and Hanna photometers 10mL Each for $66.70
Description & Specifications
Specifications
Product Type Dispenser
No. of Tests 100.0
  • Easy to dispense into round or square cuvettes
  • Each supplied with enough powder for 100 doses
  • Available for 5, 10 or 25mL sample volumes
  • Simple push button operation