HF Scientific™ Powder Pop™ Dispensers for Chlorine Testing
For testing free or total chlorine
Supplier: HF Scientific™ HF10506T
Description
- Easy to dispense into round or square cuvettes
- Each supplied with enough powder for 100 doses
- Available for 5, 10 or 25mL sample volumes
- Simple push button operation
Specifications
For testing total chlorine | |
100 | |
Dispenser |
LaMotte™, Orbeco-Hellige, YSI, palintest, tintometer and Orion photometers | |
10 mL |