HF Scientific™ Powder Pop™ Dispensers for Chlorine Testing
For testing free or total chlorine
Manufacturer:  HF Scientific™ HF10504T
 View more versions of this product
Catalog No. 15-393-154
Description
Description
  • Easy to dispense into round or square cuvettes
  • Each supplied with enough powder for 100 doses
  • Available for 5, 10 or 25mL sample volumes
  • Simple push button operation
Specifications
Specifications
Dispenser
LaMotte™, Orbeco-Hellige, YSI, palintest, tintometer and Orion photometers
10mL
For testing free chlorine
100.0
Documents