missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HF Scientific™ Micro 1000 White Light and Infrared Ratio Turbidimeters

Provide easy nephelometric measurements in less than six seconds from a 30mL sample

Manufacturer:  HF Scientific™ HF20014

 View more versions of this product

Catalog No. 15-393-95



Includes: One set of calibration standards (0.2 to 1750 NTU), operator's manual, two measuring cuvets with light shield caps, and plug-in power adapter

Description

Description

  • Read in NTU, EBC, Nephelos, FNU, and FAU
  • Autorange over entire measurement range
  • Use a quick-connect light source and feature a full-time ratio mode that automatically compensates for background color
  • Auto Alert Calibration Prompt automatically alerts operator to initiate simple, menu-driven calibration, and diagnostic software automatically monitors and detects errors

White light model (Micro 1000 WL) exceeds U.S. EPA criteria for Method 180.1; Infrared model (Micro 1000 IR) conforms to ISO 7027 standards.

Certifications

CE marked, UL listed, CSA approved.

Compliance

White light model (Micro 1000 WL) exceeds U.S. EPA criteria for Method 180.1; Infrared model (Micro 1000 IR) conforms to ISO 7027 standards.

Specifications

Specifications

±2%, ±5%, ±10%
10.75 x 10 x 3.75 in. (27 x 25 x 9.5cm)
One set of calibration standards, operator's manual, two measuring cuvets with light shield caps and plug-in power adapter
0 to 10000 NTU 0 to 2450 EBC 0 to 10000 FNU 0 to 10000 FAU
0.0001 NTU
Lithium
3.75 in.
10.75 in.
Tungsten filament lamp
30mL
0° to 50°C
2.87 lb.
10 in.
1 Year
ProCal calibration standards
Ideal and affordable tool for research as well as routine analytical measurements
Nephelometric
Micro1000 Turbidimeter
10 Years
LCD
9.5cm
27cm
<6 sec.
32° to 122°F
100/240V
1.3kg
25cm
CE, UL, CSA, US EPA
Documents