HF Scientific™ Micro 100 Laboratory Turbidimeter
Ideal for municipal water and wastewater facilities, environmental, commercial and industrial laboratories
Manufacturer:  HF Scientific™ HF20001
 View more versions of this product
Catalog No. 15-393-80
Description
Description
Ideal for municipal water and wastewater facilities, environmental, commercial and industrial laboratories
Use for research as well as routine analytical measurements.
  • Autoranging from 0 to 1000 NTU
  • Sensitive to changes as small as 0.01 NTU
  • Reusable calibration standards save time and money over mixing primary standards
  • Quick Connect Lamp Module speeds up lamp replacement and reduces maintenance downtime
Order Info
Replacement cuvettes are available: Cat. No. 15-393-5.
Specifications
Specifications
±0.01 NTU
10 x 10.7 x 4.75 in. (25.4 x 23.7 x 12.1cm)
Complete set of calibration standards (0.02, 10 and 1000 NTU), three measuring cuvettes with light shield cap, operator's manual and 120V power adapter
0 to 1000 NTU
0.01, 0.1, 1 NTU
12.1cm
25.4cm
≤6 sec.
32° to 122°F
120V
1.32kg
23.7cm
CE, US EPA
ProCal calibration standards
Ideal for municipal water and wastewater facilities, environmental, commercial and industrial laboratories
Nephelometric non-ratio
Micro 100 Laboratory Turbidimeter
4.75 in.
10 in.
Quick connect Tungsten filament or infrared
27 to 30mL
0° to 50°C
2.9 lb.
10.7 in.
1 Year
Documents