HF Scientific™ DRT-15CE Portable Turbidimeters Reusable Glass Cuvettes and Stand
For grab samples
Supplier: HF Scientific™ HF19981
Description
- Reusable 28 × 70mm glass cuvets
- Hard-tempered glass surface resists surface scratching
- Supplied with threaded light shield caps
- Cuvet stand stores up to 11 cuvets or calibration standards
Specifications
Cuvette Stand | |
DRT 15C Portable Turbidimeter | |
1 Cuvette Stand | |
1 |