missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexyldimethyloctylammonium Bromide, TCI America™

$187.92 - $556.13

Chemical Identifiers

CAS 187731-26-6
Molecular Formula C16H36BrN
Molecular Weight (g/mol) 322.375
MDL Number MFCD03093632
InChI Key KIEOLZRKTOBVMX-UHFFFAOYSA-M
PubChem CID 44630344
IUPAC Name hexyl-dimethyl-octylazanium;bromide
SMILES CCCCCCCC[N+](C)(C)CCCCCC.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09895G View Documents TCI America
H09895G
5g Each for $187.92
 
H098925G View Documents TCI America
H098925G
25g Each for $556.13
 
Specifications

Chemical Identifiers

187731-26-6
322.375
KIEOLZRKTOBVMX-UHFFFAOYSA-M
hexyl-dimethyl-octylazanium;bromide
C16H36BrN
MFCD03093632
44630344
CCCCCCCC[N+](C)(C)CCCCCC.[Br-]

Specifications

Yellow
187731-26-6
MFCD03093632
CCCCCCCC[N+](C)(C)CCCCCC.[Br-]
322.375
322.38
≥97.0% (T)
5g
C16H36BrN
KIEOLZRKTOBVMX-UHFFFAOYSA-M
hexyl-dimethyl-octylazanium;bromide
44630344
Clear Liquid at 20°C
Hexyldimethyloctylammonium Bromide
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1971

TSCA : No