Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexyl methyl ether, 98%

Chemical Identifiers

Chemical Name or Material Hexyl methyl ether
CAS 4747-07-3
Molecular Formula CH3(CH2)5OCH3
Formula Weight 116.20g/mol
MDL Number MFCD00015350
UN Number UN3271
Synonym 1-Methoxyhexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1720736

 
alfa aesar™
A17207-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Fruit-like
Quantity 500g
Chemical Name or Material Hexyl methyl ether
CAS 4747-07-3
Assay 98%
Molecular Formula CH3(CH2)5OCH3
Formula Weight 116.20g/mol
MDL Number MFCD00015350
UN Number UN3271
Synonym 1-Methoxyhexane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
TSCA TSCA

Applications
Hexyl methyl ether, finds mostly its place in masking odors and industrial fragrances. Its peculiar type of green note has tempted some perfumers to use it in Jasmin and other florals for room sprays, it can introduce very pleasant variations in Lavender and Hay fragrances.

Solubility
Soluble in ethanol fixed oils, propylene glycol. Slightly soluble in alcohol, ether, mineral oil.

Notes
This material is stable if stored under proper conditions. Avoid excessive heat and light. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.