Please login to your online account to display your discounted pricing

Hexyl Isocyanate 99%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle in Plastic Container

Manufacturer: thermo fisher scientific  411660050

 View more versions of this product

Catalog No. AC411660050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 127.19
Quantity 5g
Color Colorless to slightly yellow
Packaging Glass Bottle in Plastic Container
EINECS Number 219-763-8
TSCA TSCA
Beilstein 04, IV, 718
Infrared Spectrum Authentic
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Storage Information Product may develop some turbidity or precipitate
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Hexyl isocyanate
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H13NO
Linear Formula CH3(CH2)5NCO
MDL Number MFCD00037047
CAS 2525-62-4
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye damage.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and va
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Hexyl isocyanate
Name Note 99%
CAS 2525-62-4
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H13NO
Linear Formula CH3(CH2)5NCO
Formula Weight 127.19
MDL Number MFCD00037047
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye damage.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and va

SDS & Safety Documents