missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexyl Acrylate (stabilized with HQ) 96.0+%, TCI America™
$54.72 - $483.14
Chemical Identifiers
CAS | 2499-95-8 |
---|---|
Molecular Formula | C9H16O2 |
Molecular Weight (g/mol) | 156.225 |
MDL Number | MFCD00048881 |
InChI Key | LNMQRPPRQDGUDR-UHFFFAOYSA-N |
Synonym | 2-propenoic acid, hexyl ester, acrylic acid, hexyl ester, acrylic acid-n-hexyl ester, ageflex n-ha, ccris 7038, hexyl 2-propenoate, hexyl acrylate, hexylacrylate, n-hexyl acrylate, n-hexylacrylate |
PubChem CID | 17259 |
IUPAC Name | hexyl prop-2-enoate |
SMILES | CCCCCCOC(=O)C=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A125725ML
|
Tci America
A125725ML |
25mL |
Each for $54.72
|
|
A1257500ML
|
Tci America
A1257500ML |
500mL |
Each for $483.14
|
|
Chemical Identifiers
2499-95-8 | |
156.225 | |
LNMQRPPRQDGUDR-UHFFFAOYSA-N | |
17259 | |
CCCCCCOC(=O)C=C |
C9H16O2 | |
MFCD00048881 | |
2-propenoic acid, hexyl ester, acrylic acid, hexyl ester, acrylic acid-n-hexyl ester, ageflex n-ha, ccris 7038, hexyl 2-propenoate, hexyl acrylate, hexylacrylate, n-hexyl acrylate, n-hexylacrylate | |
hexyl prop-2-enoate |
Specifications
89°C | |
25mL | |
C9H16O2 | |
3082 | |
LNMQRPPRQDGUDR-UHFFFAOYSA-N | |
hexyl prop-2-enoate | |
17259 | |
Clear Liquid at 20°C | |
Hexyl Acrylate (stabilized with HQ) |
Yellow | |
2499-95-8 | |
MFCD00048881 | |
2-propenoic acid, hexyl ester, acrylic acid, hexyl ester, acrylic acid-n-hexyl ester, ageflex n-ha, ccris 7038, hexyl 2-propenoate, hexyl acrylate, hexylacrylate, n-hexyl acrylate, n-hexylacrylate | |
CCCCCCOC(=O)C=C | |
156.225 | |
156.23 | |
≥96.0% (GC) |
Safety and Handling
EINECSNumber : (2)-3267
RTECSNumber : AT1450000
TSCA : Yes