missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl 4-Hydroxybenzoate 98.0+%, TCI America™
Supplier: TCI America H0213500G
Specifications
| Hexyl 4-Hydroxybenzoate | |
| 53°C | |
| 171°C | |
| MFCD00016480 | |
| Hexylparaben, 4-Hydroxybenzoic Acid Hexyl Ester | |
| CCCCCCOC(=O)C1=CC=C(C=C1)O | |
| 222.284 | |
| 222.28 | |
| Crystalline Powder |
| 1083-27-8 | |
| White | |
| C13H18O3 | |
| 500 g | |
| ULULAZKOCFNOIM-UHFFFAOYSA-N | |
| hexyl 4-hydroxybenzoate | |
| 14127 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1083-27-8 | |
| 222.284 | |
| ULULAZKOCFNOIM-UHFFFAOYSA-N | |
| 14127 | |
| CCCCCCOC(=O)C1=CC=C(C=C1)O |
| C13H18O3 | |
| MFCD00016480 | |
| Hexylparaben, 4-Hydroxybenzoic Acid Hexyl Ester | |
| hexyl 4-hydroxybenzoate |
Safety and Handling
EINECSNumber : (3)-1585
RTECSNumber : DH2204000
TSCA : No