missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexetidine, mixture of stereoisomers, 97%

$98.55 - $337.73

Chemical Identifiers

CAS 141-94-6
Molecular Formula C21H45N3
Molecular Weight (g/mol) 339.612
MDL Number MFCD00010428
InChI Key DTOUUUZOYKYHEP-UHFFFAOYSA-N
Synonym hexetidine, oraldene, hexoral, glypesin, hextril, elsix, collu hextril, sterilate, sterisil, triocil
PubChem CID 3607
IUPAC Name 1,3-bis(2-ethylhexyl)-5-methyl-1,3-diazinan-5-amine
SMILES CCCCC(CC)CN1CC(CN(C1)CC(CC)CCCC)(C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1869906 View Documents Alfa Aesar™
L1869906
5g Each for $98.55
 
AAL1869914 View Documents Alfa Aesar™
L1869914
25g Each for $337.73
 
Description

Description

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

141-94-6
339.612
DTOUUUZOYKYHEP-UHFFFAOYSA-N
3607
CCCCC(CC)CN1CC(CN(C1)CC(CC)CCCC)(C)N
C21H45N3
MFCD00010428
hexetidine, oraldene, hexoral, glypesin, hextril, elsix, collu hextril, sterilate, sterisil, triocil
1,3-bis(2-ethylhexyl)-5-methyl-1,3-diazinan-5-amine

Specifications

70°C (158°F)
5g
C21H45N3
161071
hexetidine, oraldene, hexoral, glypesin, hextril, elsix, collu hextril, sterilate, sterisil, triocil
CCCCC(CC)CN1CC(CN(C1)CC(CC)CCCC)(C)N
339.612
339.61
Hexetidine, mixture of stereoisomers
160°C (0.4mmHg)
141-94-6
MFCD00010428
14,4703
DTOUUUZOYKYHEP-UHFFFAOYSA-N
1,3-bis(2-ethylhexyl)-5-methyl-1,3-diazinan-5-amine
3607
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA