missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexestrol, 98+%

Nonsteroidal synthetic estrogen

$71.85 - $71.85

Chemical Identifiers

CAS 84-16-2
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.372
MDL Number MFCD00068996
InChI Key PBBGSZCBWVPOOL-HDICACEKSA-N
Synonym hexestrol, meso-hexestrol, hexoestrol, hexanoestrol, hexestrofen, mesohexestrol, erythrohexestrol, estra-plex, cycloestrol, estrifar
PubChem CID 192197
IUPAC Name 4-[(3S,4R)-4-(4-hydroxyphenyl)hexan-3-yl]phenol
SMILES CCC(C1=CC=C(C=C1)O)C(CC)C2=CC=C(C=C2)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6088903 View Documents Alfa Aesar™
J6088903
1g Each for $71.85
 
Specifications

Chemical Identifiers

84-16-2
270.372
PBBGSZCBWVPOOL-HDICACEKSA-N
192197
CCC(C1=CC=C(C=C1)O)C(CC)C2=CC=C(C=C2)O
C18H22O2
MFCD00068996
hexestrol, meso-hexestrol, hexoestrol, hexanoestrol, hexestrofen, mesohexestrol, erythrohexestrol, estra-plex, cycloestrol, estrifar
4-[(3S,4R)-4-(4-hydroxyphenyl)hexan-3-yl]phenol

Specifications

White
1g
C18H22O2
3209460
hexestrol, meso-hexestrol, hexoestrol, hexanoestrol, hexestrofen, mesohexestrol, erythrohexestrol, estra-plex, cycloestrol, estrifar
CCC(C1=CC=C(C=C1)O)C(CC)C2=CC=C(C=C2)O
270.372
270.37
≥98%
186°C
84-16-2
MFCD00068996
14,4701
PBBGSZCBWVPOOL-HDICACEKSA-N
4-[(3S,4R)-4-(4-hydroxyphenyl)hexan-3-yl]phenol
192197
Powder
Hexestrol
Safety and Handling

Safety and Handling

GHS H Statement
H350
May cause cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : SL0560850