HexArmor™ Hercules™ 400R6E Gloves
Keeps hands safe from cuts, punctures, and scratches in high-risk situations
Manufacturer:  HexArmor™ 400R6EXL
 View more versions of this product
Catalog No. 19-130-3731D
Description
Description
Suitable for use in a wide range of industrial and animal handling applications, including waste handling/recycle processing, meat packing, lab work, and hospital waste handling and laundering. Excellent protection from bites, scratches.
  • Gauntlet cuff provides full protection for entire hand and forearm
  • Cut- and puncture-resistant1 palm and back of hand
  • Special gripping substance on palm enhances abrasion resistance and provides better grip
  • Double layered in high-risk areas
  • Fully lined interior
  • High-temp washable
1These gloves are cut and/or puncture resistant, not cut or puncture proof. Always conduct a safe test with your application to determine if the product is suited for your needs.
Specifications
Specifications
wide range of industrial and animal handling applications, including waste handling/recycle processing, meat packing, lab work and hospital waste handling and laundering
SuperFabric™
X-Large