HexArmor™ Hercules™ 400R6E Gloves
Keeps hands safe from cuts, punctures, and scratches in high-risk situations
Manufacturer: HexArmor™ 400R6EL
Description
Suitable for use in a wide range of industrial and animal handling applications, including waste handling/recycle processing, meat packing, lab work, and hospital waste handling and laundering. Excellent protection from bites, scratches.
- Gauntlet cuff provides full protection for entire hand and forearm
- Cut- and puncture-resistant1 palm and back of hand
- Special gripping substance on palm enhances abrasion resistance and provides better grip
- Double layered in high-risk areas
- Fully lined interior
- High-temp washable
1These gloves are cut and/or puncture resistant, not cut or puncture proof. Always conduct a safe test with your application to determine if the product is suited for your needs.
Specifications
wide range of industrial and animal handling applications, including waste handling/recycle processing, meat packing, lab work and hospital waste handling and laundering | |
SuperFabric™ | |
Large |