Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexarhodium hexadecacarbonyl, 98%

Chemical Identifiers

Chemical Name or Material Hexarhodium hexadecacarbonyl
CAS 28407-51-4
Molecular Formula Rh6(CO)16
Formula Weight 1065.6
MDL Number MFCD00011206
UN Number UN3466
Synonym Hexadecacarbonylhexarhodium; Rhodium carbonyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2418501

  SDS
alfa aesar™
B2418501
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2418502

  SDS
alfa aesar™
B2418502
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2418577

  SDS
alfa aesar™
B2418577
0.1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hexarhodium hexadecacarbonyl
Melting Point >300°C
Quantity 0.25g
CAS 28407-51-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
TSCA TSCA
Assay 98%
Molecular Formula Rh6(CO)16
Formula Weight 1065.6
MDL Number MFCD00011206
UN Number UN3466
Synonym Hexadecacarbonylhexarhodium; Rhodium carbonyl
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.