missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexaphenylditin, 97+%

Manufacturer:  Alfa Aesar™ 7114314

 View more versions of this product

Catalog No. AA7114314



Specifications

Specifications

Hexaphenylditin
232.5°C
25g
C36H30Sn2
UN3146
UAPQJVJCJITJET-UHFFFAOYSA-N
triphenyltin
6327129
Powder
280°C (decomposition)
1064-10-4
Characteristic
MFCD00014070
hexaphenyldistannane, hexaphenylditin, distannane, hexaphenyl, 1,1,1,2,2,2-hexaphenyldistannane, hexaphenylditin iv, hexaphenylditin iv, distannane, hexaphenyl-8ci 9ci, 1,1,1,2,2,2-hexaphenyldistannane #
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)C3=CC=CC=C3
700.056
700.02
≥97%

Chemical Identifiers

1064-10-4
700.056
UAPQJVJCJITJET-UHFFFAOYSA-N
6327129
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)C3=CC=CC=C3
C36H30Sn2
MFCD00014070
hexaphenyldistannane, hexaphenylditin, distannane, hexaphenyl, 1,1,1,2,2,2-hexaphenyldistannane, hexaphenylditin iv, hexaphenylditin iv, distannane, hexaphenyl-8ci 9ci, 1,1,1,2,2,2-hexaphenyldistannane #
triphenyltin
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

SDS Documents