Please login to your online account to display your discounted pricing

Alfa Aesar™ Hexaphenyldilead, 98%

Chemical Identifiers

Chemical Name or Material Hexaphenyldilead
CAS 3124-01-4
Molecular Formula C36H30Pb2
Linear Formula (C6H5)6Pb2
Formula Weight 877.02
MDL Number MFCD00014066
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1663706

 
alfa aesar™
A1663706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1663714

 
alfa aesar™
A1663714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1663722

 
alfa aesar™
A1663722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 877.02
Chemical Name or Material Hexaphenyldilead
Molecular Formula C36H30Pb2
MDL Number MFCD00014066
Quantity 5g
Health Hazard 3 GHS P Statement
P273-P201-P309-P310-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Linear Formula (C6H5)6Pb2
TSCA TSCA
Assay 98%
Health Hazard 1 Danger
UN Number UN3077
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Melting Point 150°C (dec.)
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
CAS 3124-01-4
Safety and Handling

Danger

GHS P Statement
P273-P201-P309-P310-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.