missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexaphenylcyclotrisiloxane, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2306106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
300°C (1mmHg) | |
∼175°C to 185°C | |
5 g | |
MFCD00023110 | |
hexaphenylcyclotrisiloxane, cyclotrisiloxane, hexaphenyl, diphenylsiloxane cyclic trimer, cyclotrisiloxane, 2,2,4,4,6,6-hexaphenyl, acmc-209kso, hexaphenyl-1,3,5,2,4,6-trioxatrisilinane, cyclotrisiloxane,2,2,4,4,6,6-hexaphenyl, 2,2,4,4,6,6-hexaphenyl-1,3,5,2,4,6 trioxatrisilinan, 2,2,4,4,6,6-hexaphenyl-1,3,5,2,4,6-trioxatrisilinane, 2,2,4,4,6,6-hexaphenyl-1,3,5,2,4,6-trioxatrisilinane # | |
C1=CC=C(C=C1)[Si]2(O[Si](O[Si](O2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7 | |
594.888 | |
594.9 | |
Moisture sensitive |
Hexaphenylcyclotrisiloxane | |
512-63-0 | |
C36H30O3Si3 | |
2957773 | |
VCYDUTCMKSROID-UHFFFAOYSA-N | |
2,2,4,4,6,6-hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane | |
68187 | |
≥98% |
Chemical Identifiers
512-63-0 | |
594.888 | |
VCYDUTCMKSROID-UHFFFAOYSA-N | |
2,2,4,4,6,6-hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane |
C36H30O3Si3 | |
MFCD00023110 | |
68187 | |
C1=CC=C(C=C1)[Si]2(O[Si](O[Si](O2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7 |
Safety and Handling
EINECSNumber : 208-145-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only