Please login to your online account to display your discounted pricing

Hexaphenylbenzene, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC165250050

 View more versions of this product

Catalog No. AC165250050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 534.7
Melting Point >300°C
Quantity 5g
Color White to off-tan
Chemical Name or Material Hexaphenylbenzene
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C42H30
Linear Formula C6(C6H5)6
MDL Number MFCD00003057
CAS 992-04-1

Specifications

Chemical Name or Material Hexaphenylbenzene
CAS 992-04-1
Molecular Formula C42H30
Linear Formula C6(C6H5)6
Formula Weight 534.7
MDL Number MFCD00003057