Please login to your online account to display your discounted pricing

Hexaphenylbenzene, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC165250010

 View more versions of this product

Catalog No. AC165250010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 534.7
Melting Point >300°C
Quantity 1g
Color White to off-tan
Packaging Glass bottle
MDL Number MFCD00003057
Chemical Name or Material Hexaphenylbenzene
CAS 992-04-1
Infrared Spectrum Authentic
Linear Formula C6(C6H5)6
Molecular Formula C42H30

Specifications

Chemical Name or Material Hexaphenylbenzene
CAS 992-04-1
Molecular Formula C42H30
Linear Formula C6(C6H5)6
Formula Weight 534.7
MDL Number MFCD00003057