Please login to your online account to display your discounted pricing

Hexaphenylbenzene, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Hexaphenylbenzene
CAS 992-04-1
Molecular Formula C42H30
Linear Formula C6(C6H5)6
Formula Weight 534.7
MDL Number MFCD00003057
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC165250050

  SDS
thermo fisher scientific
AC165250050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 534.7
Melting Point >300°C
Physical Form Powder
Quantity 5g
Color White to off-tan
Linear Formula C6(C6H5)6
Infrared Spectrum Authentic
Chemical Name or Material Hexaphenylbenzene
Molecular Formula C42H30
Packaging Glass bottle
CAS 992-04-1
MDL Number MFCD00003057