missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexaphenylbenzene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0044603
Description
Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Hexaphenylbenzene | |
>300°C | |
MFCD00003057 | |
1895903 | |
Insoluble in water. | |
C1=CC=C(C=C1)C2=C(C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7 | |
534.702 | |
534.7 |
992-04-1 | |
C42H30 | |
1 g | |
hexaphenylbenzene, benzene, hexaphenyl, 3',4',5',6'-tetraphenyl-o-terphenyl, acmc-20akri, m-terphenyl, 2',4',5',6'-tetraphenyl, ghl.pd_mitscher_leg0.890, hexaphenylbenzene 1g, 1,1':2',1-terphenyl, 3',4',5',6'-tetraphenyl, 1,2,3,4,5,6-hexaphenylbenzene | |
QBHWPVJPWQGYDS-UHFFFAOYSA-N | |
1,2,3,4,5,6-hexakis-phenylbenzene | |
70432 | |
≥98% |
Chemical Identifiers
992-04-1 | |
534.702 | |
QBHWPVJPWQGYDS-UHFFFAOYSA-N | |
70432 | |
C1=CC=C(C=C1)C2=C(C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7 |
C42H30 | |
MFCD00003057 | |
hexaphenylbenzene, benzene, hexaphenyl, 3',4',5',6'-tetraphenyl-o-terphenyl, acmc-20akri, m-terphenyl, 2',4',5',6'-tetraphenyl, ghl.pd_mitscher_leg0.890, hexaphenylbenzene 1g, 1,1':2',1-terphenyl, 3',4',5',6'-tetraphenyl, 1,2,3,4,5,6-hexaphenylbenzene | |
1,2,3,4,5,6-hexakis-phenylbenzene |
Safety and Handling
EINECSNumber : 213-591-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only