Learn More
Hexanoyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 169040050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hexanoyl chloride | |
142-61-0 | |
151.0°C to 153.0°C | |
Authentic | |
Glass bottle | |
1.4253 to 1.4273 | |
MFCD00000760 | |
02, 324 | |
15, 1765 | |
Solubility in water: hydrolysis. Other solubilities: decomposes in alcohol, soluble in ether, chloroform, ethylether and, acetone | |
CCCCCC(=O)Cl | |
134.61 | |
134.61 |
97% | |
0.9500g/mL | |
53°C | |
96.5% min. (GC) | |
C6H11ClO | |
CH3(CH2)4COCl | |
5g | |
0.95 | |
caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid | |
YWGHUJQYGPDNKT-UHFFFAOYSA-N | |
hexanoyl chloride | |
67340 | |
97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe h
GHS Signal Word: Danger
EINECSNumber : 205-549-1
TSCA : TSCA
RUO â Research Use Only