missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanoyl chloride, 97%, ACROS Organics™

Manufacturer:  Acros Organics 169045000

 View more versions of this product

Catalog No. AC169045000



Specifications

Specifications

Hexanoyl chloride
53°C
Glass bottle
0.95
Colorless to Yellow
500g
142-61-0
C6H11ClO
MFCD00000760
15, 1765
Solubility in water: hydrolysis. Other solubilities: decomposes in alcohol, soluble in ether, chloroform, ethylether and, acetone
CCCCCC(=O)Cl
134.603
134.61
97%
0.9500g/mL
Authentic
1.4253 to 1.4273
151.0°C to 153.0°C
-87.0°C
97%
97%
CH3(CH2)4COCl
02, 324
caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid
YWGHUJQYGPDNKT-UHFFFAOYSA-N
hexanoyl chloride
67340
Liquid

Chemical Identifiers

142-61-0
134.603
YWGHUJQYGPDNKT-UHFFFAOYSA-N
67340
CCCCCC(=O)Cl
C6H11ClO
MFCD00000760
caproyl chloride, n-caproyl chloride, hexanoylchloride, unii-i7ge8hg3b2, n-hexanoyl chloride, caproic acid chloride, hexanoic acid chloride, i7ge8hg3b2, caproylchloride, hexoylchlorid
hexanoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-549-1

TSCA : TSCA

SDS Documents