Learn More
Hexanoic anhydride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 186750500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexanoic anhydride | |
2051-49-2 | |
Colorless to Yellow | |
246°C to 248°C | |
Authentic | |
Glass bottle | |
1.4270 to 1.429 | |
MFCD00009509 | |
02, 324 | |
hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride | |
PKHMTIRCAFTBDS-UHFFFAOYSA-N | |
hexanoyl hexanoate | |
74918 | |
97% |
99% | |
-40°C | |
0.9260g/mL | |
130°C | |
97% | |
C12H22O3 | |
[CH3(CH2)4CO]2O | |
50 g | |
0.926 | |
Solubility in water: 72g/L. Other solubilities: soluble in ethanol and ether | |
CCCCCC(=O)OC(=O)CCCCC | |
214.3 | |
214.3 | |
Liquid |
Chemical Identifiers
2051-49-2 | |
214.3 | |
PKHMTIRCAFTBDS-UHFFFAOYSA-N | |
74918 | |
CCCCCC(=O)OC(=O)CCCCC |
C12H22O3 | |
MFCD00009509 | |
hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride | |
hexanoyl hexanoate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 218-121-4
TSCA : TSCA
RUO – Research Use Only