Learn More
Hexanoic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 120705000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexanoic acid | |
473 to 493mg KOH/g | |
Colorless to Yellow | |
202.0°C to 203.0°C | |
Authentic | |
Glass bottle | |
1.4150 to 1.4180 | |
MFCD00004421 | |
02, 321 | |
15, 1761 | |
Solubility in water: 1.1% (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCCC(=O)O | |
116.16 | |
3.23 mPa.s (20°C) | |
116.16 | |
Liquid |
142-62-1 | |
-3.0°C | |
0.9200g/mL | |
104°C | |
98.5% min. (GC) | |
C6H12O2 | |
CH3(CH2)4CO2H | |
500 g | |
0.92 | |
caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid | |
FUZZWVXGSFPDMH-UHFFFAOYSA-N | |
hexanoic acid | |
8892 | |
CHEBI:30776 | |
99% |
Chemical Identifiers
142-62-1 | |
116.16 | |
FUZZWVXGSFPDMH-UHFFFAOYSA-N | |
8892 | |
hexanoic acid |
C6H12O2 | |
MFCD00004421 | |
caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid | |
CHEBI:30776 | |
CCCCCC(=O)O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
EINECSNumber : 205-550-7
RUO – Research Use Only