Please login to your online account to display your discounted pricing

Hexanes, technical, mixture of isomers, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  210590025

 View more versions of this product

Catalog No. AC210590025

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention.
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 86.18
Quantity 2.5L
Color Clear
Merck Index 14, 4694
Beilstein 01, 142
Density 0.6590g/mL
Residue after Evaporation 0.002% max.
Linear Formula CH3(CH2)4CH3
CAS 92112-69-1
Molecular Formula C6H14
Chemical Name or Material Hexanes
MDL Number MFCD00009520
Packaging Glass bottle
Synonym Hex
CAS 110-54-3
Name Note Mixture of isomers
Health Hazard 1 Danger

Specifications

Chemical Name or Material Hexanes
Name Note Mixture of isomers
CAS 92112-69-1
CAS 110-54-3
Purity Grade Technical Grade
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Formula Weight 86.18
MDL Number MFCD00009520
Synonym Hex
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF exposed or concerned: Get medical advice/attention.

GHS H Statement
Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects.

SDS & Safety Documents