Please login to your online account to display your discounted pricing

Hexanes (Technical), Fisher Chemical


General laboratory use

Quantity 20L
Packaging Steel Pail

Manufacturer: fisher scientific company  N320

 View more versions of this product

Catalog No. N3-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Boiling Point 69°C
Melting Point −95°C
Quantity 20L
Flash Point −22°C (71.4°F)
DOT Information DOT Class 3, : Flammable Liquid
Molecular Formula C6H14
Physical Form Liquid
Color Colorless
Viscosity 0.31 mPa/s at 20°C
Vapor Pressure 160mbar at 20°C
Density 0.659g/cm³
Packaging Steel Pail
Boiling Range 30°C max., 60° to 90°C
Health Hazard 3 Emergency Overview
Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:0
ChemAlert Storage Symbol Red
Identification Pass Test
Cleanup Symbol Solvent
MDL Number MFCD02179311
Additional Information Vapor Pressure: 160mbar at 20°C
Synonym Naphtha Solvent
Health Hazard 2 DANGER!
Chemical Name or Material Hexanes
CAS 110-82-7
CAS 64742-49-0
CAS 110-54-3

Specifications

Chemical Name or Material Hexanes
CAS 110-54-3
CAS 110-82-7
CAS 64742-49-0
Purity Grade Technical Grade
MDL Number MFCD02179311
Synonym Naphtha Solvent
Accessories
 

Emergency Overview
Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents