Please login to your online account to display your discounted pricing

Hexanes, Packaged under Nitrogen, 95% n-hexane, HPLC Grade, J.T.Baker™

High purity solvents for use in analytical and preparative separations

$240.08 - $592.70

Chemical Identifiers

Chemical Name or Material Hexanes
CAS 110-54-3
Name Note Packaged under Nitrogen
Purity Grade HPLC Grade
Molecular Formula CH3(CH2)4CH3
Formula Weight 86.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002995

 
avantor performance
930403
Narrow Mouth Amber Glass Bottle 4L Each for $592.70

02002994

 
avantor performance
930402
Narrow Mouth Amber Glass Bottle 1L Each for $240.08
Description & Specifications

Specifications

Formula Weight 86.18
Molecular Formula CH3(CH2)4CH3
Name Note Packaged under Nitrogen
Purity Grade HPLC Grade
Chemical Name or Material Hexanes
Quantity 4L
Flash Point −23.00°C
Packaging Narrow Mouth Amber Glass Bottle
Assay 95% n-hexane
CAS 110-54-3
Specific Gravity 1L = 0.66kg

    For use in Liquid Chromatography (HPLC & UHPLC) & Spectrophotometry; For Laboratory, Research, or Manufacturing Use