missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanes, mixed isomers, (60+% n-hexane), Alfa Aesar™

Manufacturer:  Alfa Aesar 19353K7

 View more versions of this product

Catalog No. AA19353K7



Specifications

Specifications

Hexanes, mixed isomers
92112-69-1
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.178
CHEBI:29021
Liquid
Mixed isomers
C6H14
UN1208
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
4L

Chemical Identifiers

92112-69-1
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H315-H336

GHS P Statement
P210-P260-P201-P261-P280-P281-P301+P310-P303+P361+P353-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES

TSCA : TSCA

SDS Documents