missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanes, mixed isomers, (60+% n-hexane), Alfa Aesar™

Manufacturer:  Alfa Aesar 19353AP

 View more versions of this product

Catalog No. AA19353AP



Specifications

Specifications

Hexanes
92112-69-1
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.178
CHEBI:29021
Liquid
0.67g/mL
<−23° (−9°F)
Colorless
500mL
Mixed isomers
C6H14
UN1208
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
(≥60% n-hexane)
1.3795
65°C to 70°C
-95°C

Chemical Identifiers

92112-69-1
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H315-H336 Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES

TSCA : TSCA

SDS Documents