Learn More


Hexanes (Certified), Fisher Chemical™
Purity is guaranteed to meet published maximum limits of impurities
Manufacturer: Fisher Chemical H291200
Specifications
Hexanes | |
Vapor Pressure: 160mbar at 20°C | |
C6H14 | |
Naptha Solvent, Petroleum Naptha Solvent | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18 | |
Certified | |
Pass Test | |
0.001% max. | |
Pass Test | |
0.0003mEq/g max. | |
69°C | |
-95°C |
92112-69-1 | |
Petroleum naptha solvent | |
MFCD02179311 | |
4°C max. | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
Liquid | |
0.659g/cm³ | |
Steel Drum | |
0.005% max. | |
160mbar at 20°C | |
0.31 mPa/s at 20°C | |
Colorless | |
200L |
Chemical Identifiers
92112-69-1 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
Naptha Solvent, Petroleum Naptha Solvent | |
CHEBI:29021 | |
CCCCCC |
Safety and Handling


DANGER!
Emergency Overview
Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid