missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanes (Certified), Fisher Chemical

Purity is guaranteed to meet published maximum limits of impurities

Manufacturer:  Fisher Chemical H29120

 View more versions of this product

Catalog No. H291-20



Specifications

Specifications

Hexanes
Vapor Pressure: 160mbar at 20°C
C6H14
Naptha Solvent, Petroleum Naptha Solvent
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
Certified
Pass Test
0.001% max.
Pass Test
0.0003mEq/g max.
0.31 mPa/s at 20°C
Colorless
20L
92112-69-1
Petroleum naptha solvent
MFCD02179311
4°C max.
CCCCCC
86.18
CHEBI:29021
Liquid
0.659g/cm³
Steel Pail
0.005% max.
160mbar at 20°C
2.97g/mol
69°C
-95°C

Chemical Identifiers

92112-69-1
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
Naptha Solvent, Petroleum Naptha Solvent
CHEBI:29021
CCCCCC
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents