missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanes (Certified ACS), Fisher Chemical™

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Manufacturer:  Fisher Chemical H292500

 View more versions of this product

Catalog No. H292-500




Specifications

Specifications

Hexanes
Certified ACS
C6H14
1208
dipropyl, esani, gettysolve-b, heksan, hexan, hexanen, hexanes, hexyl hydride, n-hexane, skellysolve b
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
≥98.5%
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label.
Glass Bottle
Less-than water
Pass Test
-6.7°C (+20°F)
Colorless
500mL
92112-69-1
≥98.5 %
MFCD02179311
15, 4732
Negligible
CCCCCC
86.18
CHEBI:29021
Liquid
Certified ACS Reagent
Pass Test
0.001% max.
0.005% max.
0.0003mEq/g max.
65.5°C
-95°C

Chemical Identifiers

92112-69-1
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
dipropyl, esani, gettysolve-b, heksan, hexan, hexanen, hexanes, hexyl hydride, n-hexane, skellysolve b
CHEBI:29021
CCCCCC
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents