Please login to your online account to display your discounted pricing

Hexanes, for analysis, mixture of isomers, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC390740010

 View more versions of this product

Catalog No. AC390740010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 86.18
Physical Form Liquid
Quantity 1L
Color Clear
Density 0.6590g/mL
Purity Grade For analysis
Iron (Fe) 0.1ppm max.
Lead (Pb) 0.1ppm max.
Aromatic Substance 0.01% max. (benzene)
Linear Formula CH3(CH2)4CH3
Barium (Ba) 0.1ppm max.
Acidity 0.0002 meq/g max.
Chemical Name or Material Hexanes
Water 0.01% max.
Molecular Formula C6H14
CAS 92112-69-1
Copper (Cu) 0.02ppm max.
Magnesium (Mg) 0.1ppm max.
Packaging Glass bottle
Aluminum (Al) 0.5ppm max.
Zinc (Zn) 0.1ppm max.
Nickel (Ni) 0.02ppm max.
Residue after Evaporation 0.0005% max.
Sulfur Compounds 0.005% max.
Synonym Hex
Cadmium (Cd) 0.05ppm max.
Calcium (Ca) 0.5ppm max.
Chromium (Cr) 0.02ppm max.
Manganese (Mn) 0.02ppm max.
Substances Darkened by H2SO4 Passes Test
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H
CAS 110-54-3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Hexanes
CAS 92112-69-1
CAS 110-54-3
Purity Grade For analysis
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Formula Weight 86.18
Synonym Hex
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H

SDS & Safety Documents