Please login to your online account to display your discounted pricing

Hexanes, 99%, for spectroscopy ACS, mixture of isomers, ACROS Organics™

$64.11 - $64.11

Chemical Identifiers

Chemical Name or Material Hexanes
Name Note Mixture of isomers, for spectroscopy
CAS 92112-69-1
CAS 110-54-3
Purity Grade ACS Grade
Assay ≥98.5% (sum of 5 isomers, total hexanes, plus methylcyclopentane)
Assay Percent Range 99%
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Formula Weight 86.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC411550025

  SDS
thermo fisher scientific
AC411550025
Glass bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC411555000

  SDS
thermo fisher scientific
411555000
EcoSafPak™ Glass Bottle 500mL Each for $64.11
Description & Specifications

Specifications

Assay ≥98.5% (sum of 5 isomers, total hexanes, plus methylcyclopentane)
Purity Grade ACS Grade
Physical Form Liquid
Formula Weight 86.18
Quantity 2.5L
Color Clear
Beilstein 01, 142
Merck Index 14, 4694
EINECS Number 203-777-6
TSCA TSCA
Density 0.6590g/mL
RTECS Number MN9275000
Chemical Name or Material Hexanes
Additional Information Vapor Pressure: 160mbar at 20°C
Packaging Glass bottle
Sulfur Compounds 0.005% max.
Thiophene Passes Test
Titratable Acid 0.0003 meq/g max.
Name Note Mixture of isomers, for spectroscopy
Linear Formula CH3(CH2)4CH3
MDL Number MFCD00009520
Absorbance 0.01 max. at 280nm
Health Hazard 1 Danger
CAS 110-54-3
Residue after Evaporation 0.001% max.
Assay Percent Range 99%
Molecular Formula C6H14
CAS 92112-69-1
Synonym Hex
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For UV/VIS spectroscopy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Suspected of damaging fertility.
H